missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPC insert and body connectors, injection site

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

Brand:  Thermo Scientific™ SS00039-I

 View more versions of this product

Product Code. 17869796

  • 2025.00 € / Pack of 30

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 3 ports (1 MPC insert + cap, 1 MPC body + cap, 1 injection site). Available in 10L or 20L capacity.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors

BPC port specifications:

  • Line 1: MPC insert with 0.97cm (0.38 in.) barb, cap, and pinch clamp. Tubing: TPE 374, length: 30cm (12 in.), ID x OD: 9.5 x 16mm (0.38 x 0.63 in.)
  • Line 2: MPC body with 0.97cm (0.38 in.) barb, plug, and pinch c

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature.
Polyethylene
ASI™ 26/77
MPC connections and injection port
30/Cs.
Irradiated
5 L
TPE, 30 cm (12 in.)
4 mil. poly bag, One 4-mil. poly bag
Labtainer™
Room Temperature
32 x 31.6 cm (12.6 x 12.4 in.)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers