Learn More
LACTB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16783-1-AP-150UL
 View more versions of this product
Product Code. 16826524
  • 410.00 € / 150µL
Explore more special offers
Description
Description
Endoribonuclease; cleaves preferentially 3′ to purine-pyrimidine dinucleotide motifs in single-stranded RNA. The cleavage product contains a free 3′ -OH group. Has no activity with double-stranded RNA or DNA. Required for normal mitochondrial function and cell viability.
Specifications
Specifications
LACTB2
Polyclonal
Unconjugated
LACTB2
Beta lactamase like protein 2, CGI 83, lactamase, beta 2, LACTB2
Rabbit
Antigen Affinity Chromatography
RUO
212442, 297768, 51110
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q53H82, Q561R9, Q99KR3
LACTB2
LACTB2 Fusion Protein Ag10213
150 μL
Primary
Human, Mouse, Rat