missing translation for 'onlineSavingsMsg'
Learn More

Oxoid UK Lactic Acid 10%

A solution for addition to culture media, primarily to lower the pH

27.80 € - 84.30 €

Specifications

Content And Storage 2°C to 8°C
Country of Origin UK
Format Box
Media Type Additive
Product Type Media Supplement
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10076732
View Documents
Oxoid UK
SR0021H
10 x 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10517153
View Documents
Oxoid UK
SR0021K
10 x 5 mL
84.30 €
Pack of 10
Estimated Shipment: 26-11-2024
to see stock.
Edge
Add to basket
 
Description

Description

A solution for addition to culture media, primarily to lower the pH

Specifications

Specifications

2°C to 8°C
Box
Media Supplement
UK
Additive
Lowers pH of Culture Media
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 months

Product Identifier
  • LACTIC ACID 10%

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Lactic Acid 90%

SDS
Documents

Documents

Product Certifications