missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactic acid, lithium salt, 99%, Thermo Scientific Chemicals
23.82 € - 409.00 €
Chemical Identifiers
CAS | 867-55-0 |
---|---|
Molecular Formula | C3H5LiO3 |
Molecular Weight (g/mol) | 96.01 |
MDL Number | MFCD00065399 |
InChI Key | GKQWYZBANWAFMQ-UHFFFAOYSA-M |
Synonym | lithium lactate, lithium 2-hydroxypropanoate, lactic acid, lithium salt, dl-lactic acid lithium salt, propanoic acid, 2-hydroxy-, monolithium salt, lithium 1+ ion lactate, l-+-lactic acid lithium salt, lithotab lactate, acmc-20akmj, lithium s-lactate |
PubChem CID | 23673452 |
IUPAC Name | lithium;2-hydroxypropanoate |
SMILES | [Li+].CC(C(=O)[O-])O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10485651
|
Thermo Scientific Acros
413330050 |
5 g | Glass bottle | N/A | |||||
10301401
|
Thermo Scientific Acros
413330250 |
25 g | Glass bottle | N/A | |||||
10219433
|
Thermo Scientific Acros
413331000 |
100 g | Glass bottle | N/A | |||||
10390674
|
Thermo Scientific
413335000 |
500 g | Glass bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
867-55-0 | |
96.01 | |
GKQWYZBANWAFMQ-UHFFFAOYSA-M | |
23673452 | |
[Li+].CC(C(=O)[O-])O |
C3H5LiO3 | |
MFCD00065399 | |
lithium lactate, lithium 2-hydroxypropanoate, lactic acid, lithium salt, dl-lactic acid lithium salt, propanoic acid, 2-hydroxy-, monolithium salt, lithium 1+ ion lactate, l-+-lactic acid lithium salt, lithotab lactate, acmc-20akmj, lithium s-lactate | |
lithium;2-hydroxypropanoate |
Specifications
867-55-0 | |
White | |
Authentic | |
C3H5LiO3 | |
MFCD00065399 | |
lithium lactate, lithium 2-hydroxypropanoate, lactic acid, lithium salt, dl-lactic acid lithium salt, propanoic acid, 2-hydroxy-, monolithium salt, lithium 1+ ion lactate, l-+-lactic acid lithium salt, lithotab lactate, acmc-20akmj, lithium s-lactate | |
[Li+].CC(C(=O)[O-])O | |
96.01 | |
96.01 | |
Extra Pure | |
Lactic acid, lithium salt |
10% aq. soln., 10% aq. soln. at 0nm, 0.07 max. at 260nm | |
6 to 9 (1% soln.) | |
0.99 | |
CH3CH(OH)CO2Li | |
03, IV, 633 | |
GKQWYZBANWAFMQ-UHFFFAOYSA-M | |
lithium;2-hydroxypropanoate | |
23673452 | |
≥98.5% | |
Crystalline Powder |
Safety and Handling
Product Identifier
- Lactic acid, lithium salt
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title