missing translation for 'onlineSavingsMsg'
Learn More

Lactose, Monohydrate, Powder, N.F., J.T. Baker™

Product Code. 15567994
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 64044-51-5
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.312
InChI Key WSVLPVUVIUVCRA-ZEAJSSQNSA-N
Synonym d-lactose monohydrate, d-+-lactosemonohydrate
PubChem CID 133126686
IUPAC Name (3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O

Specifications

Packaging Poly Bottle
Quantity 2.5 kg
Specific Gravity 1L = 1.53kg
Formula Weight 360.32
Grade NF
Physical Form Powder
Chemical Name or Material Lactose, Monohydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'