missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Laemmli SDS sample buffer, non-reducing (4X)

Laemmli SDS sample buffer is used to prepare protein samples in the Laemmli SDS-PAGE system.

Brand:  Thermo Scientific Chemicals J63615.AC

61.90 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15405809

  • 62.21 € / 25mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Laemmli SDS sample buffer is used for protein sample preparation to be used in the Laemmli SDS-PAGE system, The SDS detergent denatures the proteins and subunits and gives each an overall negative charge so that each will separate based on size. The bromophenol blue serves as a dye front that runs ahead of the proteins and also serves to make it easier to see the sample during loading.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Laemmli SDS sample buffer is composed of several compounds, including 375 mM Tris-HCl (pH 6.8), 9% SDS, 50% glycerol, 9% β-mercaptoethanol, 0.03% bromophenol blue
  • The SDS compound present in the Laemmli SDS sample buffer binds noncovalently to proteins. SDS is negatively charged and can mask the intrinsic charge of the protein it binds

Application

  • Laemmli SDS sample buffer is used to denature proteins and for protein sample preparation to be used in the SDS-PAGE system
  • This sample buffer is used as an electrophoretic dye for denaturation of proteins and monitoring the front of the running gel
  • It is also used for polyacrylamide protein gel analysis
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blue
Sample Loading Buffer
Laemmli SDS-PAGE
4X
Liquid
25 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY18K550 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberJ63615.AD,J63615.AC
Lot NumberW06K548 Certificate TypeCertificate of Analysis Date10/10/2024 Catalog NumberJ63615.AD,J63615.AC
Lot NumberR17K504 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog NumberJ63615.AD,J63615.AC
Lot NumberR17K505 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog NumberJ63615.AD,J63615.AC
Lot NumberQ19K507 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ63615.AD,J63615.AC
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Laemmli SDS sample buffer, non-reducing (4X) > 25mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only