Learn More
Thermo Scientific Chemicals Laemmli SDS sample buffer, non-reducing (6X)
Laemmli SDS sample buffer is used to prepare protein samples in the Laemmli SDS-PAGE system.
88.14 € - 138.69 €
Specifications
Color | Blue |
---|---|
Concentration | 6X |
Product Type | Sample Loading Buffer |
Form | Liquid |
For Use With (Application) | Laemmli SDS-PAGE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15413399
|
Thermo Scientific Chemicals
J60660.AC |
25 mL |
88.14 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15423399
|
Thermo Scientific Chemicals
J60660.AD |
50 mL |
138.69 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Laemmli SDS sample buffer, non-reducing (6X) is an electrophoretic dye for denaturation of proteins and monitoring the front of running gel. Composition of this buffer is similar to the reducing buffer minus mercaptoethanol. This product is ideal for polyacrylamide protein gel analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Blue | |
Sample Loading Buffer | |
Laemmli SDS-PAGE |
6X | |
Liquid |
Safety and Handling
- Laemmli SDS sample buffer, non-reducing (6X)
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 1,2,3-Propanetriol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.