missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Laemmli SDS Sample Buffer with Pyronin Y, non-reducing, 4X
For protein sample preparation to be used in the Laemmli SDS-PAGE system
35.00 € - 57.00 €
Specifications
Color | Pink |
---|---|
Concentration | 4X |
Product Type | Sample Loading Buffer |
Form | Liquid |
For Use With (Application) | Laemmli SDS-PAGE |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Pink | |
Sample Loading Buffer | |
Laemmli SDS-PAGE |
4X | |
Liquid |