Learn More
Brady™ Laminated Polyester ADR 1.5 1 Explosive Substances Transport Signs
Brand:  Brady™ 223557
 View more versions of this product
Product Code. 11760204
  • 14.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
ADR 1.5-297*297-B7541
Laminated Polyester
Permanent Acrylic
0.099 mm
-40 °C-110 °C
1.5 1
Gloss
1 Ea.
297 mm
297.00 mm
Diamond
Black on Orange