Learn More
Brady™ Laminated Polyester ADR 1.5 1 Explosive Substances Transport Signs
Brand:  Brady™ 223595
 View more versions of this product
Product Code. 11790234
  • 441.00 € / 1 roll
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
ADR 1.5RL-100*100-B7541
Laminated Polyester
Permanent Acrylic
0.099 mm
-40 °C-110 °C
1.5 1
Gloss
250/Pk.
100 mm
100.00 mm
Diamond
Black on Orange