Learn More
Brady™ Laminated Polyester: ISO Safety Sign - No access for people with active implanted cardiac devices
Sign compliant to ISO 7010.
Brand:  Brady™ 822739
 View more versions of this product
Product Code. 15654617
  • 5.70 € / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Laminated polyester
  • For indoor and outdoor use
Specifications
Specifications
ISO 7010
Gloss
2/Pk.
50 mm
0.099 mm
Round
Black on White
PIC P007-Dia 050-PE-SHEET/2
Polyester
Permanent Acrylic
Indoor and Outdoor
-40°C to 110°C
Global