Learn More
Brady™ Laminated Polyester: ISO Safety Sign - No open flame; Fire, open ignition source and smoking prohibited
Sign compliant to ISO 7010.
Brand:  Brady™ 822146
 View more versions of this product
Product Code. 15674577
  • 4.85 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Laminated polyester
  • For indoor and outdoor use
Specifications
Specifications
ISO 7010
Gloss
1 Ea.
200 mm
0.099 mm
Round
Black on White
PIC P003-Dia 200-PE-CRD/1
Polyester
Permanent Acrylic
Indoor and Outdoor
-40°C to 110°C
Global