missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Laminated Polyester: ISO Safety Sign - Warning: Biological hazard
Sign compliant to ISO 7010.
4.75 € - 462.00 €
Specifications
Finish | Gloss |
---|---|
Material | Polyester |
Adhesive Type | Permanent Acrylic |
For Use With (Application) | Indoor and Outdoor |
Thickness | 0.099 mm |
Product Code | Brand | Width (Metric) | Height (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Height (Metric) | Quantity | Price | Quantity & Availability | ||||
15654707
|
Brady™
827649 |
50 mm | 43 mm | 7/Pk. |
7.18 €
Pack of 7 |
In Stock
Log in to see stock. |
||||
15548471
|
Brady™
827651 |
100 mm | 87 mm | 3/Pk. |
12.48 €
Pack of 3 |
In Stock
Log in to see stock. ![]() |
||||
15342759
|
Brady™
827650 |
100 mm | 87 mm | 250/Pk. |
462.00 €
1 roll |
In Stock
Log in to see stock. |
||||
15367667
|
Brady™
827652 |
200 mm | 173 mm | 1 Ea. |
4.75 €
Each |
In Stock
Log in to see stock. |
||||
15664707
|
Brady™
827653 |
315 mm | 273 mm | 1 Ea. |
7.65 €
Each |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||
Description
- Laminated polyester
- For indoor and outdoor use
Specifications
Gloss | |
Permanent Acrylic | |
0.099 mm | |
Triangular | |
Black on Yellow |
Polyester | |
Indoor and Outdoor | |
-40°C to 110°C | |
Global |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ Laminated Polyester: ISO Safety Sign - Warning: Biological hazard