Learn More
Brady™ Laminated Polyester Stickers for the Transport of Infectious Substances
Brand:  Brady™ 257535
 View more versions of this product
Product Code. 10080284
  • 6.95 € / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
ADR 6.2-200*200-B7541
Laminated Polyester
Permanent Acrylic
0.099 mm
-40 °C-110 °C
6
Gloss
1 Ea.
200 mm
200.00 mm
Rhomboid
Black on White