Learn More
Brady™ Laminated Polyester Stickers for the Transport of Infectious Substances
Brand:  Brady™ 257536
 View more versions of this product
Product Code. 11744884
  • 14.00 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
ADR 6.2-297*297-B7541
Laminated Polyester
Permanent Acrylic
0.099 mm
-40 °C-110 °C
6
Gloss
1 Ea.
297 mm
297.00 mm
Rhomboid
Black on White