Learn More
Brady™ Laminated Polyester Transport Sign
Brand:  Brady™ 257525
 View more versions of this product
Product Code. 10162454
  • 6.95 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Gloss
1 Ea.
Laminated Polyester
200 mm
200 mm
Diamond
Transport Sign
ADR 8-200*200-B7541
Polyester
Permanent Acrylic
Pipe markers and safety signs
0.024 mm
-40°C to 110°C
Corrosive Substance 8
Black on White