Learn More
Brady™ Laminated Polyester Transport Sign
Brand:  Brady™ 223586
 View more versions of this product
Product Code. 11700234
  • 441.00 € / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Gloss
250/Roll
Laminated Polyester
100 mm
100 mm
Diamond
Transport Sign
ADR 9RL-100*100-B7541
Polyester
Permanent
Pipe markers and safety signs
0.024 mm
-40°C to 110°C
Dangerous Substance 9
Clear