Learn More
Brady™ Laminated Polyester Transport Sign
Brand:  Brady™ 223601
 View more versions of this product
Product Code. 11750244
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
ISO 7010, RoHS compliant using EU Directive 2002/95/EC, ROHS Environmental Compliance
Polyester
Permanent
Pipe markers and safety signs
0.024 mm
-40 °C-110 °C
Non-flammable, non-toxic gas 2
White on Green
Gloss
250/Roll
Laminated Polyester
100 mm
100 mm
Diamond
Transport Sign