Learn More
UVP Lamp XX-20BLB bench brackets included aluminum 220-240V 50/60Hz length 610mm supplied with 2 x 365nm BLB 20W tubes UVP
Brand:  UVP 95-0045-05
Product Code. 12802979
  • 697.00 € / Each
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
UVP Lamp XX-20BLB bench brackets included aluminum 220-240V 50/60Hz length 610mm supplied with 2 x 365nm BLB 20W tubes UVP >