Learn More
Lanthanum aluminum oxide, 99.9% (metals basis), Thermo Scientific Chemicals
86.40 € - 251.00 €
Chemical Identifiers
CAS | 12003-65-5 |
---|---|
MDL Number | MFCD00144550 |
Synonym | Lanthanum aluminate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11333648
|
Thermo Scientific Alfa Aesar
039250.06 |
5 g |
86.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343648
|
Thermo Scientific Alfa Aesar
039250.14 |
25 g |
251.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in epitaxial thin films, non-epitaxial thin films and substrate. Mainly used in making optical glass parts and optical fibers for electronic industry. Also often used in ceramic industry and catalysts. Due to its dielectric properties it is also posses properties for low-loss microwave and dielectric resonance applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12003-65-5 | |
MFCD00144550 | |
Lanthanum aluminate |
Specifications
12003-65-5 | |
MFCD00144550 | |
Soluble in phosphorous acid (at >150°C). Insoluble in mineral acid ( at 25°C) and water. | |
99.9% | |
(metals basis) |
Powder | |
Lanthanum aluminate | |
213.88 | |
Odorless | |
Lanthanum aluminum oxide |
Safety and Handling
missing translation for 'einecsNumber' : 234-433-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only