missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Aluminum Oxide Substrate, 10 x 10 x 0.5mm, Polished Two Sides, 100 Orientation, Thermo Scientific™

100 Orientation

111.00 € - 470.00 €

Chemical Identifiers

CAS 12003-65-5
Molecular Formula LaAlO3
MDL Number MFCD00144550
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11344148
SDS View Documents
Thermo Scientific Alfa Aesar
041434.KT
1 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11354148
SDS View Documents
Thermo Scientific Alfa Aesar
041434.KU
5 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

12003-65-5
LaAlO3
MFCD00144550

Specifications

12003-65-5
Plate
MFCD00144550
Lanthanum Aluminum Oxide Substrate, Polished Two Sides
Brown-Yellow
LaAlO3
Odorless
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.