missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum chloride, 99.9%, Thermo Scientific Chemicals

93.26 € - 1101.48 €

Chemical Identifiers

CAS 10099-58-8
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00011068
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10287660
SDS View Documents
Thermo Scientific Acros
315230100
10 g Glass bottle
93.26 €
10g
Please to purchase this item. Need a web account? Register with us today!
10404545
SDS View Documents
Thermo Scientific Acros
315230500
50 g Glass bottle
313.56 €
50g
Please to purchase this item. Need a web account? Register with us today!
13448518
SDS View Documents
Thermo Scientific Acros
315232500
250 g Glass bottle
1101.48 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10099-58-8
245.26
ICAKDTKJOYSXGC-UHFFFAOYSA-K
[Cl-].[Cl-].[Cl-].[La+3]
Cl3La
MFCD00011068
lanthanum(3+) trichloride

Specifications

1000.0°C
10099-58-8
Powder
Cl3La
MFCD00011068
Solubility in water: soluble
[Cl-].[Cl-].[Cl-].[La+3]
245.26
99.9%
860.0°C
White
99.9%
LaCl3
15,5414
ICAKDTKJOYSXGC-UHFFFAOYSA-K
lanthanum(3+) trichloride
245.26
Lanthanum chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Causes serious eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 233-237-5

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.