missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum chloride, hydrated, 64.5-70% LaCl3, Thermo Scientific Chemicals

Product Code. 10144360
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20211-76-1
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00149753
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]

Specifications

Color White
Physical Form Adhering Crystals
Quantity 100 g
Linear Formula LaCl3·xH2O
Merck Index 10,5193
Solubility Information Solubility in water: soluble
Formula Weight 245.27
Percent Purity 64.5 to 70% LaCl3
Packaging Glass bottle
Chemical Name or Material Lanthanum chloride, hydrated
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lanthanum chloride, hydrated
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'