missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Chloride 10% Solution, for AAS, Fisher Chemical™

Product Code. 10050250
Click to view available options
Quantity:
250 mL
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

MDL Number 149756

Specifications

pH 1
Physical Form Liquid
Quantity 250 mL
Assay Percent Range ≥9.5 and ≤10.5% w/v
Packaging Amber glass bottle
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lanthanum chloride solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.