missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Chloride 10% Solution, for AAS, Fisher Chemical™

Product Code. p-7073255 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 10050250

Brand: Fisher Chemical L/0460/07

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

MDL Number 149756

Specifications

pH 1
Physical Form Liquid
Quantity 250 mL
Assay Percent Range ≥9.5 and ≤10.5% w/v
Packaging Amber glass bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.