Learn More
Lanthanum(III) 2,4-pentanedionate hydrate, 99.9% (REO), Thermo Scientific Chemicals
118.59 € - 354.77 €
Chemical Identifiers
CAS | 64424-12-0 |
---|---|
Molecular Formula | C15H21LaO6 |
Molecular Weight (g/mol) | 436.23 |
MDL Number | MFCD00149057 |
InChI Key | UUYNRMJIFVOQCR-UHFFFAOYSA-N |
Synonym | lanthanum acetylacetonate hydrate min |
PubChem CID | 132984033 |
IUPAC Name | (Z)-4-hydroxypent-3-en-2-one;lanthanum;dihydrate |
SMILES | [La+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11342288
|
Thermo Scientific Alfa Aesar
013194.14 |
25 g |
118.59 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11352288
|
Thermo Scientific Alfa Aesar
013194.22 |
100 g |
354.77 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a catalysts, laboratory chemicals & for manufacture of chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64424-12-0 | |
436.23 | |
UUYNRMJIFVOQCR-UHFFFAOYSA-N | |
132984033 | |
[La+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
C15H21LaO6 | |
MFCD00149057 | |
lanthanum acetylacetonate hydrate min | |
(Z)-4-hydroxypent-3-en-2-one;lanthanum;dihydrate |
Specifications
143°C | |
White to Yellow | |
C15H21LaO6 | |
lanthanum acetylacetonate hydrate min | |
UUYNRMJIFVOQCR-UHFFFAOYSA-N | |
(Z)-4-hydroxypent-3-en-2-one;lanthanum;dihydrate | |
132984033 | |
99.9% | |
(REO) |
64424-12-0 | |
Powder | |
MFCD00149057 | |
Insoluble in water. | |
[La+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
436.23 | |
436.24 (Anhydrous) | |
Hygroscopic | |
Lanthanum(III) 2,4-pentanedionate hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 238-187-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only