missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) acetate sesquihydrate, 99.9% (REO), Thermo Scientific Chemicals

60.80 € - 237.18 €

Chemical Identifiers

CAS 25721-92-0
Molecular Formula C12H24La2O15
Molecular Weight (g/mol) 686.12
MDL Number MFCD00013037
InChI Key RBPQRWPSWRTKEC-UHFFFAOYSA-H
IUPAC Name dilanthanum(3+) hexaacetate trihydrate
SMILES O.O.O.[La+3].[La+3].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11347118
View Documents
Thermo Scientific Alfa Aesar
015299.22
100 g
60.80 €
100g
Please to purchase this item. Need a web account? Register with us today!
11357118
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
015299.36
500 g
237.18 €
500g
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

Lanthanum(III) acetate sesquihydrate is used in specialty glass and in water treatment. It is used in the preparation of phenolate-bridged dilanthanum(III) complex, which is considered as a model for metalloproteins in applied chemistry. Further, it is used to prepare nanoparticulate porous lanthanum oxyfluoride (LaOF) films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Especificaciones

Identificadores de productos químicos

25721-92-0
686.12
RBPQRWPSWRTKEC-UHFFFAOYSA-H
O.O.O.[La+3].[La+3].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
C12H24La2O15
MFCD00013037
dilanthanum(3+) hexaacetate trihydrate

Especificaciones

25721-92-0
C12H24La2O15
Soluble in water.
O.O.O.[La+3].[La+3].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
686.12
99.9%
(REO)
Lump/Powder
MFCD00013037
RBPQRWPSWRTKEC-UHFFFAOYSA-H
dilanthanum(3+) hexaacetate trihydrate
343.05 (316.03 Anhydrous)
Hygroscopic
Lanthanum(III) acetate sesquihydrate
Videos
Seguridad y manipulación

Seguridad y manipulación

missing translation for 'einecsNumber' : 213-034-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documentos

Documentos

Certificados

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado