missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11348697
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a scintillation material in certain application, as a source in the synthesis of lanthanum & its derivatives CsNaLa6Br14N2 and La3Br6N which contain rare bitetrahedral structural units..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13536-79-3
Molecular Formula Br3La
Molecular Weight (g/mol) 378.62
MDL Number MFCD00016161
InChI Key XKUYOJZZLGFZTC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) tribromide
SMILES [Br-].[Br-].[Br-].[La+3]

Specifications

Physical Form Beads
Quantity 2 g
Solubility Information Soluble in water.
Formula Weight 378.62
Percent Purity 99.9%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lanthanum(III) bromide, Ultra dry

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'