missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) carbonate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11337198
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used in pharmaceuticals & used in medicine as a phosphate binder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54451-24-0
Molecular Formula C3La2O9
Molecular Weight (g/mol) 457.84
MDL Number MFCD00011069
InChI Key NZPIUJUFIFZSPW-UHFFFAOYSA-H
IUPAC Name dilanthanum(3+) tricarbonate
SMILES [La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Physical Form Powder
Quantity 500 g
Merck Index 14,5364
Solubility Information Insoluble in water.
Formula Weight 457.85 (Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Density 2.6 g/mL to 2.7 g/mL
Assay (REO)
Chemical Name or Material Lanthanum(III) carbonate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'