Learn More
Lanthanum(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
Description
Lanthanum chloride solutions are used in various water treatment applications. It is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
Boiling Point | 1,000°C |
Melting Point | 860°C |
Color | White |
Physical Form | Beads |
Quantity | 10 g |
UN Number | UN3077 |
Merck Index | 14,5363 |
Solubility Information | Soluble in water. |
Formula Weight | 245.27 |
Percent Purity | 99.9% |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.