missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Chemical Identifiers

CAS 10099-58-8
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00011068
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11318607
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035731.06
5 g N/A
N/A
11308607
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035731.14
25 g N/A
N/A
11398597
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035731.22
100 g N/A
N/A
Description

Description

Lanthanum chloride solutions are used in various water treatment applications. It is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

10099-58-8
245.26
ICAKDTKJOYSXGC-UHFFFAOYSA-K
[Cl-].[Cl-].[Cl-].[La+3]
Cl3La
MFCD00011068
lanthanum(3+) trichloride

Specifications

1,000°C
10099-58-8
Granules
MFCD00011068
14,5363
ICAKDTKJOYSXGC-UHFFFAOYSA-K
lanthanum(3+) trichloride
245.27
Odorless
Hygroscopic
(REO)
860°C
White
Cl3La
UN3077
Soluble in water.
[Cl-].[Cl-].[Cl-].[La+3]
245.26
99.99%
Ampule
3.842 g/mL
Lanthanum(III) chloride, Ultra dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H290-H317
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.

P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c

H290-H317-H318

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III

missing translation for 'einecsNumber' : 233-237-5

missing translation for 'rtecsNumber' : OE4375000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only