Learn More
Lanthanum(III) hydroxide, 99.95% (REO), Thermo Scientific Chemicals
Chemical Identifiers
CAS | 14507-19-8 |
---|---|
Molecular Formula | La(OH)3 |
MDL Number | MFCD00016160 |
Synonym | lanthanum hydroxide, la oh 3, lanthanum hydroxide van, lanthanum 3+ trihydroxide, acmc-1c707, lanthanum 3+ ion trihydroxide, lanthanum iii hydroxide trace rare earth metals basis 50g |
Description
Lanthanum(III) hydroxide is used in three way catalyst, glass, ceramic and electronic industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14507-19-8 | |
La(OH)3 | |
MFCD00016160 | |
lanthanum hydroxide, la oh 3, lanthanum hydroxide van, lanthanum 3+ trihydroxide, acmc-1c707, lanthanum 3+ ion trihydroxide, lanthanum iii hydroxide trace rare earth metals basis 50g |
Specifications
(decomposition) | |
White | |
La(OH)3 | |
lanthanum hydroxide, la oh 3, lanthanum hydroxide van, lanthanum 3+ trihydroxide, acmc-1c707, lanthanum 3+ ion trihydroxide, lanthanum iii hydroxide trace rare earth metals basis 50g | |
189.93 | |
Odorless | |
Lanthanum (III) hydroxide |
14507-19-8 | |
Powder | |
MFCD00016160 | |
Insoluble in water. Soluble in strong acids. | |
99.95% | |
(REO) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 238-510-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only