missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Chemical Identifiers

CAS 13813-22-4
Molecular Formula I3La
Molecular Weight (g/mol) 519.62
MDL Number MFCD00049470
InChI Key KYKBXWMMXCGRBA-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) triiodide
SMILES [I-].[I-].[I-].[La+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11358697
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035801.03
1 g N/A
N/A
11368697
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035801.06
5 g N/A
N/A
Description

Description

Lanthanum(III) iodide is used as a catalyst in the transesterification of N-acyloxazolidinones with alcohol gives the corresponding esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

13813-22-4
519.62
KYKBXWMMXCGRBA-UHFFFAOYSA-K
[I-].[I-].[I-].[La+3]
I3La
MFCD00049470
lanthanum(3+) triiodide

Specifications

772°C
Beads
MFCD00049470
KYKBXWMMXCGRBA-UHFFFAOYSA-K
lanthanum(3+) triiodide
519.62
Odorless
Hygroscopic
Lanthanum(III) iodide, Ultra dry
13813-22-4
I3La
Soluble in water.
[I-].[I-].[I-].[La+3]
519.62
99.9%
Ampule
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 237-465-6

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only