missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) iodide, ultra dry, 99.995+% (metals basis), Thermo Scientific Chemicals

135.68 € - 497.48 €

Chemical Identifiers

CAS 13813-22-4
Molecular Formula I3La
Molecular Weight (g/mol) 519.62
MDL Number MFCD00049470
InChI Key KYKBXWMMXCGRBA-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) triiodide
SMILES [I-].[I-].[I-].[La+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11415310
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044548.03
1 g
135.68 €
1g
Please to purchase this item. Need a web account? Register with us today!
11425310
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044548.06
5 g
497.48 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

13813-22-4
519.62
KYKBXWMMXCGRBA-UHFFFAOYSA-K
[I-].[I-].[I-].[La+3]
I3La
MFCD00049470
lanthanum(3+) triiodide

Specifications

772°C
Beads
MFCD00049470
[I-].[I-].[I-].[La+3]
519.62
≥99.995%
Moisture sensitive
Lanthanum (III) iodide, Ultra dry
13813-22-4
I3La
KYKBXWMMXCGRBA-UHFFFAOYSA-K
lanthanum(3+) triiodide
519.62
Ampule
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 237-465-6

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only