missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11347647
Click to view available options
Quantity:
100 g
500 g

Lanthanum(III) nitrate is utilized as a starting material for the electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. It is used as a catalyst for the chemoselective preparation of cyclic and acyclic dithioacetals. It is used to prepare lanthanum aluminate (LAO) thin films, LaF3 nanocrystals, and bis(indolyl) methanes from indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10277-43-7
Molecular Formula H12LaN3O15
Molecular Weight (g/mol) 433.01
MDL Number MFCD00149751
InChI Key GJKFIJKSBFYMQK-UHFFFAOYSA-N
Synonym lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate
PubChem CID 16211478
IUPAC Name lanthanum(3+);trinitrate;hexahydrate
SMILES O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 40°C
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.9% (REO)
UN Number UN1477
Merck Index 14,5363
Solubility Information Soluble in water,alcohol and acids.
Formula Weight 433.01 (324.92 Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Lanthanum(III) nitrate hexahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lanthanum(III) nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Lanthanum(III) nitrate hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.