missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) nitrate hexahydrate, Honeywell Fluka™
Puriss. p.a.,≥99.0% (titration)
181.87 € - 843.02 €
Chemical Identifiers
CAS | 10277-43-7 |
---|---|
Molecular Formula | H12LaN3O15 |
Molecular Weight (g/mol) | 433.01 |
MDL Number | MFCD00149751 |
InChI Key | GJKFIJKSBFYMQK-UHFFFAOYSA-N |
Synonym | lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate |
PubChem CID | 16211478 |
IUPAC Name | lanthanum(3+);trinitrate;hexahydrate |
SMILES | O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15636040
|
Honeywell-Fluka
61520-100G |
100 g |
181.87 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15646040
|
Honeywell-Fluka
61520-500G |
500 g |
843.02 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
10277-43-7 | |
433.01 | |
GJKFIJKSBFYMQK-UHFFFAOYSA-N | |
16211478 | |
O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
H12LaN3O15 | |
MFCD00149751 | |
lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate | |
lanthanum(3+);trinitrate;hexahydrate |
Specifications
10277-43-7 | |
La(NO3)3 · 6H2O | |
UN1477 | |
GJKFIJKSBFYMQK-UHFFFAOYSA-N | |
lanthanum(3+);trinitrate;hexahydrate | |
16211478 | |
≥99% (titration) |
H12LaN3O15 | |
MFCD00149751 | |
lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate | |
O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
433.01 | |
433.01g/mol | |
Lanthanum(III) nitrate hexahydrate |
Safety and Handling
P220-P261-P305 + P351 + P338
missing translation for 'rtecsNumber' : OE5250000