Learn More
Lanthanum(III) nitrate hydrate, 99%, water ca 22-25%, Thermo Scientific™
35.00 € - 135.00 €
Chemical Identifiers
CAS | 100587-94-8 |
---|---|
Molecular Formula | LaN3O9 |
Molecular Weight (g/mol) | 324.92 |
MDL Number | MFCD00011067 |
InChI Key | FYDKNKUEBJQCCN-UHFFFAOYSA-N |
Synonym | lanthanum iii nitrate hydrate, lanthanum nitrate hydrate, nitric acid,lanthanum 3+ salt, hydrate 9ci, acmc-1c39v, la.3no3.h2o, lanthanum nitrate-water 1/3/1, lanthanum 3+ hydrate trinitrate, lanthanum 3+ ion hydrate trinitrate, lanthanum nitrate, releasing agent solution, specpure, lanthanum iii nitrate hydrate trace metals basis |
PubChem CID | 45051674 |
SMILES | [La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsLanthanum(III) nitrate hydrate serves as a raw material for the electrochemical synthesis of LaMnO4, which is a thin film coating on stainless steel substrate. It is also used in preparation of La2O2S. Further, it is used in phosphors, fluorescent display tubes and heat transfer measurement.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong reducing agents and strong acids.
Chemical Identifiers
100587-94-8 | |
324.92 | |
FYDKNKUEBJQCCN-UHFFFAOYSA-N | |
45051674 |
LaN3O9 | |
MFCD00011067 | |
lanthanum iii nitrate hydrate, lanthanum nitrate hydrate, nitric acid,lanthanum 3+ salt, hydrate 9ci, acmc-1c39v, la.3no3.h2o, lanthanum nitrate-water 1/3/1, lanthanum 3+ hydrate trinitrate, lanthanum 3+ ion hydrate trinitrate, lanthanum nitrate, releasing agent solution, specpure, lanthanum iii nitrate hydrate trace metals basis | |
[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
40°C | |
LaN3O9 | |
MFCD00011067 | |
14,5363 | |
Soluble in water. | |
[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
324.92 | |
324.92 (Anhydrous) | |
water ≈22 to 25% |
100587-94-8 | |
La(NO3)3·xH2O | |
UN1477 | |
lanthanum iii nitrate hydrate, lanthanum nitrate hydrate, nitric acid,lanthanum 3+ salt, hydrate 9ci, acmc-1c39v, la.3no3.h2o, lanthanum nitrate-water 1/3/1, lanthanum 3+ hydrate trinitrate, lanthanum 3+ ion hydrate trinitrate, lanthanum nitrate, releasing agent solution, specpure, lanthanum iii nitrate hydrate trace metals basis | |
FYDKNKUEBJQCCN-UHFFFAOYSA-N | |
lanthanum(3+) trinitrate | |
45051674 | |
99% | |
Lanthanum(III) nitrate hydrate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 233-238-0
missing translation for 'rtecsNumber' : OE5075000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures