missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) oxalate decahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11392288
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 537-03-1
Molecular Formula C6La2O12
Molecular Weight (g/mol) 541.865
MDL Number MFCD00150540
InChI Key OXHNIMPTBAKYRS-UHFFFAOYSA-H
Synonym lanthanum oxalate, lanthanum iii oxalate, acmc-1bafu, lanthanum 3+ ; oxalate, dilanthanum 3+ trioxalate ion, dilanthanum 3+ trioxalate, lanthanum iii oxalate decahydrate, reacton, ethanedioic acid,lanthanum 3+ salt,hydrate 3:2:9, lanthanum, m-ethanedioato 2--ko1,ko2':ko1',ko2 bis ethanedioato 2--ko1,ko2 di
PubChem CID 164654
IUPAC Name lanthanum(3+);oxalate
SMILES C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].[La+3].[La+3]

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 50 g
Linear Formula La2(C2O4)3·10H2O
UN Number UN3288
Formula Weight 722.02 (541.87 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lanthanum(III) oxalate decahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'