missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) oxalate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Lanthanum Oxalate is mainly applied in specialty glass, phosphors, water treatment and catalyst. This material use in glass, ceramics, paper, textile, coating, fine and specialty chemicals. In petroleum cracking catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | (decomposition) |
Physical Form | Powder |
Quantity | 100 g |
Linear Formula | La2(C2O4)3·xH2O |
UN Number | UN3288 |
Solubility Information | Soluble in water. Soluble in strong mineral acids. |
Formula Weight | 541.87 (Anhydrous) |
Sensitivity | Hygroscopic |
Chemical Name or Material | Lanthanum (III) oxalate hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'