missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11398778
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

For vacuum deposition

Lanthanum(III) oxide is used for vacuum deposition. It is also acts as a precursor to LAMOX fast ion conductors. It is also employed in the manufacture of special alloys, optical glass and refractory materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-81-8
MDL Number MFCD00011071

Specifications

Boiling Point 4200°C
Melting Point 2307°C
Physical Form Sintered Lump
Quantity 250 g
Merck Index 14,5363
Solubility Information Insoluble in water; Soluble in dilute mineral acids
Formula Weight 325.82
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 6.51 g/mL
Assay (metals basis)
Chemical Name or Material Lanthanum(III) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'