missing translation for 'onlineSavingsMsg'
Learn More
Lanthanum(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11366348

Lanthanum(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11366348
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11366348

Brand: Thermo Scientific Alfa Aesar 011272.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lanthanum(III) oxide is used as a precursor to LAMOX fast ion conductors and superconductors. It is also used in the manufacture of optical glasses, piezoelectric and thermoelectric material. It is also useful for X-ray imaging intensifying screens, phosphors as well as dielectric and conductive ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-81-8
MDL Number MFCD00011071
Synonym Lanthanum trioxide, Lanthanum sesquioxide

Specifications

Boiling Point 4200°C
Melting Point 2307°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9% (REO)
Molecular Formula La2O3
Solubility Information Insoluble in water but soluble in dilute acid.
Formula Weight 325.82
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder/lump
Density 6.51 g/cm3 at 20°C
Chemical Name or Material Lanthanum(III) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lanthanum(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.