missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

50.50 € - 478.00 €

Chemical Identifiers

CAS 1312-81-8
Molecular Formula La2O3
MDL Number MFCD00011071
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11396328
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011264.18
50 g
50.50 €
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11306338
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011264.30
250 g
167.00 €
250g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11386328
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011264.A1
1 kg
478.00 €
1kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lanthanum(III) oxide is used as a precursor to LAMOX fast ion conductors and superconductors. It is also used in the manufacturing of optical glasses, piezoelectric and thermoelectric material. It is also useful for X-ray imaging intensifying screens, phosphors as well as dielectric and conductive ceramics. In addition, it is used as a catalyst to effect certain organic transformations. It is one of the active ingredients and finds application in thermoelectric and piezoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1312-81-8
La2O3
MFCD00011071

Specifications

4200°C
1312-81-8
Powder
La2O3
Insoluble in water. Soluble in dilute acid.
Odorless
6.51 g/cm3 at 20°C
2307°C
White
99.99% (REO)
MFCD00011071
325.82
Hygroscopic
Lanthanum(III) oxide, Rare Earth Oxides Content
Videos
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

missing translation for 'einecsNumber' : 215-200-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only