missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) oxide, REacton™, 99.999% (REO), Thermo Scientific Chemicals

194.97 € - 933.65 €

Chemical Identifiers

CAS 1312-81-8
Molecular Formula La2O3
MDL Number MFCD00011071
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11336338
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011265.18
50 g
194.97 €
50g
Please to purchase this item. Need a web account? Register with us today!
11326338
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011265.30
250 g
539.69 €
250g
Please to purchase this item. Need a web account? Register with us today!
11316338
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011265.A1
1 kg
933.65 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lanthanum(III) oxide is used as a precursor to LAMOX fast ion conductors and superconductors. It is also used in the manufacture of optical glasses, piezoelectric and thermoelectric material. It is also useful for X-ray imaging intensifying screens, phosphors as well as dielectric and conductive ceramics. In addition, it is used as a catalyst to effect certain organic transformations. It is one of the active ingredients and finds application in thermoelectric and piezoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1312-81-8
La2O3
MFCD00011071

Specifications

4200°C
1312-81-8
99.999% (REO)
MFCD00011071
Insoluble in water. Soluble in dilute acid.
99.999%
Hygroscopic
(REO)
2307°C
Powder
La2O3
14,5363
325.82
Reacton™
6.51 g/mL
Lanthanum(III) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a

Warning

missing translation for 'einecsNumber' : 215-200-5

missing translation for 'rtecsNumber' : OE5330000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only