missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) perchlorate hexahydrate, Thermo Scientific Chemicals

49.04 € - 148.74 €

Chemical Identifiers

CAS 14017-46-0
Molecular Formula Cl3H12LaO18
Molecular Weight (g/mol) 545.33
MDL Number MFCD00150245
InChI Key DCEUOMQVLPJQAI-UHFFFAOYSA-K
Synonym lanthanum perchlorate, lanthanum 3+ perchlorate, perchloric acid, lanthanum 3+ salt, perchloric acid, lanthanum 3+ salt 3:1, lanthanum 3+ triperchlorate, lanthanum iii perchlorate solution
PubChem CID 166977
IUPAC Name lanthanum(3+);triperchlorate
SMILES O.O.O.O.O.O.[La+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11361748
SDS View Documents
Thermo Scientific Alfa Aesar
011641.14
25 g
49.04 €
25g
Please to purchase this item. Need a web account? Register with us today!
11351748
SDS View Documents
Thermo Scientific Alfa Aesar
011641.22
100 g
148.74 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lanthanum(III) perchlorate hexahydrate participates in the preparation of the Schiff base complexes of lanthanide (III) ions. When hydrated lanthanum perchlorate in ethanolic solution is treated with an ethanolic solution of terpyridine, [La(terpy)3](CIO4)3 is deposited as very small crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14017-46-0
545.33
DCEUOMQVLPJQAI-UHFFFAOYSA-K
166977
O.O.O.O.O.O.[La+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Cl3H12LaO18
MFCD00150245
lanthanum perchlorate, lanthanum 3+ perchlorate, perchloric acid, lanthanum 3+ salt, perchloric acid, lanthanum 3+ salt 3:1, lanthanum 3+ triperchlorate, lanthanum iii perchlorate solution
lanthanum(3+);triperchlorate

Specifications

14017-46-0
Cl3H12LaO18
UN1481
Easily soluble in water.
O.O.O.O.O.O.[La+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
545.33
545.35 (437.26 Anhydrous)
Hygroscopic
Crystalline
MFCD00150245
lanthanum perchlorate, lanthanum 3+ perchlorate, perchloric acid, lanthanum 3+ salt, perchloric acid, lanthanum 3+ salt 3:1, lanthanum 3+ triperchlorate, lanthanum iii perchlorate solution
DCEUOMQVLPJQAI-UHFFFAOYSA-K
lanthanum(3+);triperchlorate
166977
Odorless
Lanthanum(III) perchlorate hexahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.

P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c

H272-H315-H319

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 237-835-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.