missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) sulfate, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

113.57 € - 449.24 €

Chemical Identifiers

CAS 10099-60-2
Molecular Formula La2O12S3
Molecular Weight (g/mol) 565.979
MDL Number MFCD00011070
InChI Key VQEHIYWBGOJJDM-UHFFFAOYSA-H
Synonym lanthanum sulfate, sulfuric acid, lanthanum 3+ salt 3:2, dilanthanum 3+ trisulphate, lanthanum iii sulfate 2:3, dilanthanum 3+ trisulfate, lanthansulfat, lanthanum sulphate, lanthanum 3+ trisulfate, 2la.3so4, lanthanum iii sulfatehydrate
PubChem CID 159259
IUPAC Name lanthanum(3+);trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[La+3].[La+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11483820
View Documents Promotion Details
Thermo Scientific Alfa Aesar
089760.18
50 g
113.57 €
50g
Please to purchase this item. Need a web account? Register with us today!
11493820
View Documents Promotion Details
Thermo Scientific Alfa Aesar
089760.30
250 g
449.24 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used for atomic weight determinations for lanthanum. Also used as catalyst and in electronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10099-60-2
565.979
VQEHIYWBGOJJDM-UHFFFAOYSA-H
159259
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[La+3].[La+3]
La2O12S3
MFCD00011070
lanthanum sulfate, sulfuric acid, lanthanum 3+ salt 3:2, dilanthanum 3+ trisulphate, lanthanum iii sulfate 2:3, dilanthanum 3+ trisulfate, lanthansulfat, lanthanum sulphate, lanthanum 3+ trisulfate, 2la.3so4, lanthanum iii sulfatehydrate
lanthanum(3+);trisulfate

Specifications

1150°C (decomposition)
White
La2O12S3
14,5363
Soluble in water.
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[La+3].[La+3]
565.979
566
Odorless
3.60 g/mL
Lanthanum (III) sulfate, Anhydrous
10099-60-2
Powder
MFCD00011070
lanthanum sulfate, sulfuric acid, lanthanum 3+ salt 3:2, dilanthanum 3+ trisulphate, lanthanum iii sulfate 2:3, dilanthanum 3+ trisulfate, lanthansulfat, lanthanum sulphate, lanthanum 3+ trisulfate, 2la.3so4, lanthanum iii sulfatehydrate
VQEHIYWBGOJJDM-UHFFFAOYSA-H
lanthanum(3+);trisulfate
159259
99.9%
Hygroscopic
(REO)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 233-239-6

missing translation for 'rtecsNumber' : OE5400000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only