missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Product Code. 10776442
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52093-26-2
Molecular Formula C3F9LaO9S3
Molecular Weight (g/mol) 586.1
MDL Number MFCD00077560
InChI Key WGJJZRVGLPOKQT-UHFFFAOYSA-K
Synonym lanthanum iii trifluoromethanesulfonate, lanthanum triflate, lanthanum trifluoromethanesulfonate, la otf 3, lanthanum iii triflate, lanthanum tris trifluoromethanesulfonate, lanthanum 3+ tritriflate, lanthanum 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid lanthanum salt, tris trifluoromethanesulfonato lanthanum
PubChem CID 2733938
IUPAC Name lanthanum(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[La+3]

Specifications

Color White to Beige
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 99%
Solubility Information Solubility in water: soluble in water
Formula Weight 586.1
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.7
Density 1.7000g/mL
Chemical Name or Material Lanthanum(III) trifluoromethanesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Lanthanum(III) trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'