missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

30.25 € - 87.33 €

Chemical Identifiers

CAS 52093-26-2
Molecular Formula C3F9LaO9S3
Molecular Weight (g/mol) 586.1
MDL Number MFCD00077560
InChI Key WGJJZRVGLPOKQT-UHFFFAOYSA-K
Synonym lanthanum iii trifluoromethanesulfonate, lanthanum triflate, lanthanum trifluoromethanesulfonate, la otf 3, lanthanum iii triflate, lanthanum tris trifluoromethanesulfonate, lanthanum 3+ tritriflate, lanthanum 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid lanthanum salt, tris trifluoromethanesulfonato lanthanum
PubChem CID 2733938
IUPAC Name lanthanum(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[La+3]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10776442
SDS View Documents
Thermo Scientific Acros
343910010
1 g Glass bottle
30.25 €
1g
Please to purchase this item. Need a web account? Register with us today!
10606194
SDS View Documents
Thermo Scientific Acros
343910050
5 g Glass bottle
87.33 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

52093-26-2
586.1
WGJJZRVGLPOKQT-UHFFFAOYSA-K
2733938
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[La+3]
C3F9LaO9S3
MFCD00077560
lanthanum iii trifluoromethanesulfonate, lanthanum triflate, lanthanum trifluoromethanesulfonate, la otf 3, lanthanum iii triflate, lanthanum tris trifluoromethanesulfonate, lanthanum 3+ tritriflate, lanthanum 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid lanthanum salt, tris trifluoromethanesulfonato lanthanum
lanthanum(3+);trifluoromethanesulfonate

Specifications

52093-26-2
Crystalline Powder
C3F9LaO9S3
lanthanum iii trifluoromethanesulfonate, lanthanum triflate, lanthanum trifluoromethanesulfonate, la otf 3, lanthanum iii triflate, lanthanum tris trifluoromethanesulfonate, lanthanum 3+ tritriflate, lanthanum 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid lanthanum salt, tris trifluoromethanesulfonato lanthanum
WGJJZRVGLPOKQT-UHFFFAOYSA-K
lanthanum(3+);trifluoromethanesulfonate
2733938
99%
1.7
Lanthanum(III) trifluoromethanesulfonate
White to Beige
99%
MFCD00077560
Solubility in water: soluble in water
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[La+3]
586.1
586.1
Authentic
1.7000g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Product Identifier
  • Lanthanum(III) trifluoromethanesulfonate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.