missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum Nitrate solution EU Pharmacopoeia, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Specifications
Specifications
Concentration | 100μg/mL |
CAS | 33820-53-0 |
Packaging | Bottle |
Quantity | 1000 mL |
Molecular Formula | C15H23N3O4 |
InChI Key | NEKOXWSIMFDGMA-UHFFFAOYSA-N |
SMILES | CCCN(CCC)C1=C(C=C(C=C1[N+](=O)[O-])C(C)C)[N+](=O)[O-] |
IUPAC Name | 2,6-dinitro-4-propan-2-yl-N,N-dipropylaniline |
Molecular Weight (g/mol) | 309.366 |
PubChem CID | 36606 |
Show More |
Safety and Handling
Product Identifier
- Lanthanum nitrate solution. European Pharmacopoeia Ref: 1048001
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P391-Collect spillage.
- MIXTURE LIST-Contains : anthanum nitrate, hexahydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction