missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum pieces, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Silver-White |
Appearance | Silvery-white pieces |
Density | 6.17g/cm3 at 20°C |
Assay Percent Range | 99.9% (REO) |
Quantity | 25 g |
Packaging | Under Mineral Oil |
Chemical Name or Material | Lanthanum, Rare Earth Oxides Content |
Safety and Handling

Product Identifier
- Lanthanum pieces
- Danger
- Flammable solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
- H228-Flammable solid.
- H261-In contact with water releases flammable gases.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use to extinguish.
- P402+P404-Store in a dry place. Store in a closed container.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lanthanum pieces, 99.9% (REO), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction