missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL

Product Code. p-4408560
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
This item is not returnable. View return policy

Product Code. 11341449

Brand: Thermo Scientific Alfa Aesar 013870.AD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1312-81-8 | La2O3 in 5% HNO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-81-8, 7697-37-2, 7732-18-5
Molecular Formula La2O3 in 5% HNO3
MDL Number MFCD00011066

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Color Colorless
Quantity 50 mL
Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Lanthanum(III) oxide: 0.12%; Water: 94.88%
Chemical Name or Material Lanthanum

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.